siRNA Details
| Gene Symbol: Cdkn1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031762.3 | NP_113950.2 | 2 | 254 | ||
| XM_006237539.2 | XP_006237601.1 | 1 | 707 | |||
| GenBank mRNA | AF015194.1 | - | 1 | 178 | ||
| AY623024.1 | - | 1 | 600 | |||
| AY623040.1 | - | 2 | 254 | |||
| D83792.1 | - | 1 | 540 | |||
| D86924.1 | - | 1 | 178 | |||

Cdkn1b
Genomic Map