siRNA Details
| Gene Symbol: Rxrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031765.1 | NP_113953.1 | 5 | 939 | ||
| XM_006250212.2 | XP_006250274.1 | 4 | 1083 | |||
| GenBank mRNA | AF059312.1 | - | 1 | 67 | ||
| AJ223083.1 | - | 4 | 474 | |||
| BC091363.1 | - | 5 | 939 | 7319081 | ||
| Gene Symbol: LOC102553986 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250213.2 | XP_006250275.1 | 1 | 1544 | ||

Rxrg
Genomic Map