siRNA Details
| Gene Symbol: Rxrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031765.1 | NP_113953.1 | 8 | 1310 | ||
| XM_006250212.2 | XP_006250274.1 | 7 | 1454 | |||
| GenBank mRNA | AF016387.1 | - | 1 | 14 | ||
| AJ223083.1 | - | 7 | 845 | |||
| BC091363.1 | - | 8 | 1310 | 7319081 | ||

Rxrg
Genomic Map