siRNA Details
| Gene Symbol: Gnb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031770.3 | NP_113958.1 | 10 | 927 | ||
| XM_008766344.1 | XP_008764566.1 | 12 | 1563 | |||
| GenBank mRNA | AF001953.1 | - | 10 | 927 | ||
| AY552803.1 | - | 10 | 927 | |||
| BC089221.1 | - | 10 | 941 | 7320619 | ||
| DQ120491.1 | - | 10 | 934 | |||
| DQ120492.1 | - | 10 | 934 | |||

Gnb5
Genomic Map