siRNA Details
| Gene Symbol: Gda | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031776.2 | NP_113964.2 | 10 | 997 | ||
| GenBank mRNA | AF026472.2 | - | 10 | 1054 | ||
| AF245172.1 | - | 10 | 997 | |||
| FQ233257.1 | - | 10 | 1020 | |||
| FQ233523.1 | - | 10 | 1025 | |||

Gda
Genomic Map