siRNA Details
| Gene Symbol: Atp6ap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031785.1 | NP_113973.1 | 7 | 888 | ||
| GenBank mRNA | AF035387.1 | - | 7 | 888 | ||
| BC070937.1 | - | 7 | 888 | 7099117 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp6ap1| Gene Symbol: Atp6ap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031785.1 | NP_113973.1 | 7 | 888 | ||
| GenBank mRNA | AF035387.1 | - | 7 | 888 | ||
| BC070937.1 | - | 7 | 888 | 7099117 | ||