siRNA Details
| Gene Symbol: Cit | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029911.1 | NP_001025082.1 | 40 | 5321 | ||
| XM_006249469.2 | XP_006249531.1 | 42 | 5389 | |||
| XM_006249470.2 | XP_006249532.1 | 42 | 5344 | |||
| XM_006249471.2 | XP_006249533.1 | 41 | 5344 | |||
| XM_006249472.2 | XP_006249534.1 | 41 | 5299 | |||
| XM_006249477.2 | XP_006249539.1 | 32 | 4577 | |||
| XM_008769269.1 | XP_008767491.1 | 42 | 5389 | |||
| XM_008769270.1 | XP_008767492.1 | 42 | 5345 | |||
| XM_008769271.1 | XP_008767493.1 | 41 | 5415 | |||
| GenBank mRNA | AF039218.1 | - | 32 | 4515 | ||

Cit
Genomic Map