siRNA Details
| Gene Symbol: Ugcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031795.2 | NP_113983.1 | 3 | 339 | ||
| GenBank mRNA | AF047707.1 | - | 3 | 401 | ||
| AJ224156.1 | - | 3 | 325 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ugcg| Gene Symbol: Ugcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031795.2 | NP_113983.1 | 3 | 339 | ||
| GenBank mRNA | AF047707.1 | - | 3 | 401 | ||
| AJ224156.1 | - | 3 | 325 | |||