siRNA Details
| Gene Symbol: Taldo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031811.3 | NP_113999.2 | 6 | 734 | ||
| GenBank mRNA | AF069306.1 | - | 6 | 720 | ||
| BC059126.1 | - | 6 | 738 | 6889440 | ||
| FQ213939.1 | - | 6 | 746 | |||
| FQ219168.1 | - | 6 | 760 | |||

Taldo1
Genomic Map