siRNA Details
Gene Symbol: Cd164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031812.1 | NP_114000.1 | 4, 5 | 370 | ||
GenBank mRNA | AJ238574.1 | - | 4, 5 | 370 | ||
BC062064.1 | - | 4, 5 | 373 | 5599253 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cd164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031812.1 | NP_114000.1 | 4, 5 | 370 | ||
GenBank mRNA | AJ238574.1 | - | 4, 5 | 370 | ||
BC062064.1 | - | 4, 5 | 373 | 5599253 |