siRNA Details
| Gene Symbol: Rbbp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031816.1 | NP_114004.1 | 12 | 1516 | ||
| GenBank mRNA | AF090306.1 | - | 12 | 1516 | ||
| BC062012.1 | - | 12 | 1425 | 5621292 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbbp7| Gene Symbol: Rbbp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031816.1 | NP_114004.1 | 12 | 1516 | ||
| GenBank mRNA | AF090306.1 | - | 12 | 1516 | ||
| BC062012.1 | - | 12 | 1425 | 5621292 | ||