siRNA Details
| Gene Symbol: Fat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031819.2 | NP_114007.1 | 2 | 2166 | ||
| GenBank mRNA | AF100960.1 | - | 2 | 2168 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fat1| Gene Symbol: Fat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031819.2 | NP_114007.1 | 2 | 2166 | ||
| GenBank mRNA | AF100960.1 | - | 2 | 2168 | ||