siRNA Details
| Gene Symbol: Fat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031819.2 | NP_114007.1 | 10 | 5608 | ||
| XM_008771269.1 | XP_008769491.1 | 8 | 2235 | |||
| GenBank mRNA | AF100960.1 | - | 10 | 5610 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fat1| Gene Symbol: Fat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031819.2 | NP_114007.1 | 10 | 5608 | ||
| XM_008771269.1 | XP_008769491.1 | 8 | 2235 | |||
| GenBank mRNA | AF100960.1 | - | 10 | 5610 | ||