siRNA Details
Gene Symbol: Plk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031821.1 | NP_114009.1 | 13 | 1938 | ||
GenBank mRNA | AF136583.1 | - | 13 | 1938 | ||
BC070878.1 | - | 13 | 1922 | 7096750 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Plk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031821.1 | NP_114009.1 | 13 | 1938 | ||
GenBank mRNA | AF136583.1 | - | 13 | 1938 | ||
BC070878.1 | - | 13 | 1922 | 7096750 |