siRNA Details
| Gene Symbol: Ncoa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031822.1 | NP_114010.1 | 9 | 1659 | ||
| GenBank mRNA | AF136943.1 | - | 9 | 1659 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ncoa2| Gene Symbol: Ncoa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031822.1 | NP_114010.1 | 9 | 1659 | ||
| GenBank mRNA | AF136943.1 | - | 9 | 1659 | ||