siRNA Details
Gene Symbol: Flot2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270800.1 | NP_001257729.1 | 4, 5 | 459 | ||
NM_001270801.1 | NP_001257730.1 | 4, 5 | 468 | |||
NM_031830.2 | NP_114018.1 | 4, 5 | 459 | |||
XM_006246952.1 | XP_006247014.1 | 5, 6 | 1080 | |||
XM_008768119.1 | XP_008766341.1 | 3, 4 | 446 | |||
GenBank mRNA | AF023302.1 | - | 4, 5 | 438 | ||
AF023303.1 | - | 4, 5 | 438 | |||
AF023304.1 | - | 3, 4 | 356 | |||
BC083550.1 | - | 3, 4 | 335 | 7133999 | ||
CB581049.1 | - | 4, 5 | 459 | |||
U64999.1 | - | 1, 2 | 311 |
