siRNA Details
Gene Symbol: Flot2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270800.1 | NP_001257729.1 | 7 | 695 | ||
NM_001270801.1 | NP_001257730.1 | 7 | 704 | |||
NM_031830.2 | NP_114018.1 | 7 | 695 | |||
XM_006246952.1 | XP_006247014.1 | 8 | 1316 | |||
XM_008768119.1 | XP_008766341.1 | 6 | 682 | |||
GenBank mRNA | AF023302.1 | - | 7 | 674 | ||
AF023303.1 | - | 7 | 674 | |||
AF023304.1 | - | 6 | 592 | |||
BC083550.1 | - | 6 | 571 | 7133999 | ||
U64999.1 | - | 4 | 547 |
