siRNA Details
| Gene Symbol: Rtn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031831.1 | NP_114019.1 | 4 | 3227 | ||
| XM_006251605.2 | XP_006251667.1 | 4 | 3413 | |||
| XM_006251606.1 | XP_006251668.1 | 4 | 3413 | |||
| XM_006251608.1 | XP_006251670.1 | 2 | 1004 | |||
| XM_006251609.2 | XP_006251671.1 | 2 | 714 | |||
| XM_008770468.1 | XP_008768690.1 | 3 | 4814 | |||
| GenBank mRNA | AF051335.1 | - | 2 | 139 | ||
| AF132045.1 | - | 3 | 1323 | |||
| AF132046.1 | - | 2 | 803 | |||
| AJ242961.1 | - | 4 | 3227 | |||
| AJ242962.1 | - | 2 | 754 | |||
| AJ242963.1 | - | 2 | 200 | |||
| AY164740.1 | - | 2 | 754 | |||
| AY164741.1 | - | 3 | 1323 | |||
| BC070879.1 | - | 2 | 939 | |||
| BC097936.1 | - | 2 | 759 | |||
| BC161841.1 | - | 2 | 760 | |||

Rtn4
Genomic Map