siRNA Details
| Gene Symbol: Rtn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031831.1 | NP_114019.1 | 5 | 3512 | ||
| XM_006251605.2 | XP_006251667.1 | 5 | 3698 | |||
| XM_006251608.1 | XP_006251670.1 | 3 | 1289 | |||
| XM_006251609.2 | XP_006251671.1 | 3 | 999 | |||
| XM_008770468.1 | XP_008768690.1 | 4 | 5099 | |||
| GenBank mRNA | AF051335.1 | - | 3 | 424 | ||
| AF132045.1 | - | 4 | 1608 | |||
| AF132046.1 | - | 3 | 1088 | |||
| AJ242961.1 | - | 5 | 3512 | |||
| AJ242962.1 | - | 3 | 1039 | |||
| AJ242963.1 | - | 3 | 485 | |||
| AY164740.1 | - | 3 | 1039 | |||
| AY164741.1 | - | 4 | 1608 | |||
| BC070879.1 | - | 3 | 1224 | |||
| BC097936.1 | - | 3 | 1044 | |||
| BC161841.1 | - | 3 | 1045 | |||
| FQ213608.1 | - | 1 | 84 | |||
| FQ228713.1 | - | 2 | 222 | |||
| FQ235341.1 | - | 2 | 185 | |||

Rtn4
Genomic Map