siRNA Details
Gene Symbol: Rtn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031831.1 | NP_114019.1 | 3 | 2674 | ||
XM_006251605.2 | XP_006251667.1 | 3 | 2860 | |||
XM_006251606.1 | XP_006251668.1 | 3 | 2860 | |||
XM_008770468.1 | XP_008768690.1 | 2 | 4261 | |||
GenBank mRNA | AJ242961.1 | - | 3 | 2674 |
