siRNA Details
| Gene Symbol: Sept9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113497.1 | NP_001106969.1 | 465 | |||
| NM_031837.2 | NP_114025.2 | 1353 | ||||
| NM_176856.2 | NP_789826.2 | 1472 | ||||
| GenBank mRNA | AF170253.1 | - | 1473 | |||
| AF173899.1 | - | 1352 | ||||
| AF180525.1 | - | 4, 5 | 632 | |||
| AF180526.1 | - | 453 | ||||
| BC166602.1 | - | 1018 | ||||
| CF977797.1 | - | 4, 5 | 491 | |||

Sept9
Genomic Map