siRNA Details
| Gene Symbol: Psmd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031978.1 | NP_114184.1 | 14 | 1588 | ||
| GenBank mRNA | AJ006340.1 | - | 14 | 1588 | ||
| BC087654.1 | - | 14 | 1673 | |||
| BC105883.1 | - | 14 | 1634 | |||
| BC128734.1 | - | 14 | 1633 | |||
| BC166420.1 | - | 14 | 1629 | |||
| FQ218653.1 | - | 3 | 205 | |||
| FQ234100.1 | - | 3 | 190 | |||

Psmd1
Genomic Map