siRNA Details
| Gene Symbol: Trpv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031982.1 | NP_114188.1 | 16 | 2510 | ||
| XM_008767882.1 | XP_008766104.1 | 15 | 2517 | |||
| GenBank mRNA | AB040873.1 | - | 15 | 2430 | ||
| AB041029.1 | - | 14 | 2250 | |||
| AF029310.1 | - | 16 | 2510 | |||
| AF158248.2 | - | 10 | 1843 | |||
| LC008303.1 | - | 11 | 1750 | |||

Trpv1
Genomic Map