siRNA Details
| Gene Symbol: Ache | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172009.1 | NP_742006.1 | 1 | 678 | ||
| GenBank mRNA | BC094521.1 | - | 773 | 7315968 | ||
| S50879.1 | - | 1 | 678 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ache| Gene Symbol: Ache | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172009.1 | NP_742006.1 | 1 | 678 | ||
| GenBank mRNA | BC094521.1 | - | 773 | 7315968 | ||
| S50879.1 | - | 1 | 678 | |||