siRNA Details
| Gene Symbol: Ache | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172009.1 | NP_742006.1 | 3 | 1632 | ||
| GenBank mRNA | AY555735.1 | - | 1 | 79 | ||
| BC094521.1 | - | 1727 | 7315968 | |||
| S50879.1 | - | 3 | 1632 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ache| Gene Symbol: Ache | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172009.1 | NP_742006.1 | 3 | 1632 | ||
| GenBank mRNA | AY555735.1 | - | 1 | 79 | ||
| BC094521.1 | - | 1727 | 7315968 | |||
| S50879.1 | - | 3 | 1632 | |||