siRNA Details
Gene Symbol: Dclk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053343.3 | NP_445795.1 | 11 | 1103 | ||
XM_008761004.1 | XP_008759226.1 | 11 | 3005 | |||
XM_008761005.1 | XP_008759227.1 | 11 | 2031 | |||
XM_008761006.1 | XP_008759228.1 | 11 | 3008 | |||
XM_008761007.1 | XP_008759229.1 | 11 | 1115 | |||
GenBank mRNA | CB586027.1 | - | 4 | 281 | ||
FQ211799.1 | - | 11 | 1090 | |||
U78857.1 | - | 11 | 1093 |
