siRNA Details
| Gene Symbol: Smarcd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031983.2 | NP_114189.1 | 8 | 1144 | ||
| GenBank mRNA | AB003504.1 | - | 8 | 874 | ||
| BC062063.1 | - | 8 | 1144 | 5599204 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smarcd2| Gene Symbol: Smarcd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031983.2 | NP_114189.1 | 8 | 1144 | ||
| GenBank mRNA | AB003504.1 | - | 8 | 874 | ||
| BC062063.1 | - | 8 | 1144 | 5599204 | ||