siRNA Details
| Gene Symbol: Nde1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053347.1 | NP_445799.1 | 4 | 508 | ||
| XM_006245849.2 | XP_006245911.1 | 5 | 625 | |||
| XM_006245850.2 | XP_006245912.1 | 6 | 657 | |||
| XM_006245851.1 | XP_006245913.1 | 5 | 625 | |||
| XM_008767443.1 | XP_008765665.1 | 6 | 678 | |||
| XM_008767444.1 | XP_008765666.1 | 6 | 678 | |||
| XM_008767445.1 | XP_008765667.1 | 6 | 678 | |||
| GenBank mRNA | AF240463.1 | - | 4 | 508 | ||
| BC081759.1 | - | 4 | 520 | 7190483 | ||

Nde1
Genomic Map