siRNA Details
| Gene Symbol: Ltbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191561.1 | NP_001178490.1 | 20 | 2952 | ||
| XM_006230906.2 | XP_006230968.1 | 20 | 3004 | |||
| XM_006230908.2 | XP_006230970.1 | 20 | 2953 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ltbp3| Gene Symbol: Ltbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191561.1 | NP_001178490.1 | 20 | 2952 | ||
| XM_006230906.2 | XP_006230968.1 | 20 | 3004 | |||
| XM_006230908.2 | XP_006230970.1 | 20 | 2953 | |||