siRNA Details
| Gene Symbol: Calb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031984.2 | NP_114190.1 | 2, 3 | 264 | ||
| GenBank mRNA | BC081764.1 | - | 2, 3 | 247 | 7110962 | |
| CK479935.1 | - | 2, 3 | 265 | |||
| FQ214326.1 | - | 2, 3 | 281 | |||
| X04280.1 | - | 2, 3 | 279 | |||

Calb1
Genomic Map