siRNA Details
| Gene Symbol: Crot | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031987.1 | NP_114193.1 | 4 | 412 | ||
| XM_006236004.2 | XP_006236066.1 | 4 | 572 | |||
| XM_006236005.2 | XP_006236067.1 | 5 | 1902 | |||
| GenBank mRNA | BC074004.1 | - | 4 | 432 | 7098081 | |
| J02844.1 | - | 853 | ||||
| U26033.1 | - | 4 | 412 | |||

Crot
Genomic Map