siRNA Details
| Gene Symbol: Kalrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032062.2 | NP_114451.2 | 57 | 8101 | ||
| GenBank mRNA | AF232669.1 | - | 56 | 8071 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kalrn| Gene Symbol: Kalrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032062.2 | NP_114451.2 | 57 | 8101 | ||
| GenBank mRNA | AF232669.1 | - | 56 | 8071 | ||