siRNA Details
| Gene Symbol: Grip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032069.1 | NP_114458.1 | 9 | 1299 | ||
| GenBank mRNA | DQ227256.1 | - | 1169 | |||
| U88572.1 | - | 9 | 1299 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grip1| Gene Symbol: Grip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032069.1 | NP_114458.1 | 9 | 1299 | ||
| GenBank mRNA | DQ227256.1 | - | 1169 | |||
| U88572.1 | - | 9 | 1299 | |||