siRNA Details
| Gene Symbol: Grip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032069.1 | NP_114458.1 | 11 | 1690 | ||
| GenBank mRNA | AY437398.1 | - | 677 | |||
| DQ227255.1 | - | 2 | 330 | |||
| DQ227256.1 | - | 1560 | ||||
| U88572.1 | - | 11 | 1690 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grip1| Gene Symbol: Grip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032069.1 | NP_114458.1 | 11 | 1690 | ||
| GenBank mRNA | AY437398.1 | - | 677 | |||
| DQ227255.1 | - | 2 | 330 | |||
| DQ227256.1 | - | 1560 | ||||
| U88572.1 | - | 11 | 1690 | |||