siRNA Details
| Gene Symbol: Ryr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191043.1 | NP_001177972.1 | 89 | 13540 | ||
| NM_032078.2 | NP_114467.1 | 90 | 13464 | |||
| GenBank mRNA | AB204523.1 | - | 89 | 13540 | ||
| EU346200.2 | - | 90 | 13464 | |||
| U95157.1 | - | 4 | 535 | |||

Ryr2
Genomic Map