siRNA Details
| Gene Symbol: Pnliprp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032081.1 | NP_114470.1 | 4 | 292 | ||
| GenBank mRNA | AB072249.1 | - | 2 | 65 | ||
| FQ232772.1 | - | 4 | 291 | |||
| X61925.1 | - | 4 | 292 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pnliprp1| Gene Symbol: Pnliprp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032081.1 | NP_114470.1 | 4 | 292 | ||
| GenBank mRNA | AB072249.1 | - | 2 | 65 | ||
| FQ232772.1 | - | 4 | 291 | |||
| X61925.1 | - | 4 | 292 | |||