siRNA Details
| Gene Symbol: Hao2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032082.2 | NP_114471.1 | 6, 7 | 967 | ||
| XM_006233022.2 | XP_006233084.1 | 7, 8 | 1132 | |||
| XM_006233024.2 | XP_006233086.1 | 7, 8 | 1123 | |||
| XM_006233025.2 | XP_006233087.1 | 6, 7 | 991 | |||
| GenBank mRNA | BC078781.1 | - | 5, 6 | 883 | 7132928 | |
| X67156.1 | - | 6, 7 | 968 | |||

Hao2
Genomic Map