siRNA Details
| Gene Symbol: Ssbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053358.1 | NP_445810.1 | 10, 11 | 1017 | ||
| XM_006238534.2 | XP_006238596.1 | 11, 12 | 1156 | |||
| XM_006238535.2 | XP_006238597.1 | 11, 12 | 1130 | |||
| XM_006238538.2 | XP_006238600.1 | 10, 11 | 1072 | |||
| XM_008763905.1 | XP_008762127.1 | 11, 12 | 2807 | |||
| XM_008763906.1 | XP_008762128.1 | 11, 12 | 2780 | |||
| XM_008763909.1 | XP_008762131.1 | 10, 11 | 2696 | |||
| GenBank mRNA | AF121893.1 | - | 10, 11 | 1017 | ||
| AY724490.1 | - | 11, 12 | 798 | |||
| BC107666.1 | - | 10, 11 | 1001 | 7936459 | ||
| FQ230168.1 | - | 5, 6 | 295 | |||

Ssbp3
Genomic Map