siRNA Details
| Gene Symbol: Abcd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033352.1 | NP_203503.1 | 1 | 995 | ||
| GenBank mRNA | AF131294.1 | - | 1 | 995 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abcd2| Gene Symbol: Abcd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033352.1 | NP_203503.1 | 1 | 995 | ||
| GenBank mRNA | AF131294.1 | - | 1 | 995 | ||