siRNA Details
| Gene Symbol: Dffb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053362.1 | NP_445814.1 | 7 | 941 | ||
| GenBank mRNA | AF136598.1 | - | 7 | 941 | ||
| BC129091.1 | - | 1182 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dffb| Gene Symbol: Dffb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053362.1 | NP_445814.1 | 7 | 941 | ||
| GenBank mRNA | AF136598.1 | - | 7 | 941 | ||
| BC129091.1 | - | 1182 | ||||