siRNA Details
| Gene Symbol: Clcn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053363.2 | NP_445815.2 | 7 | 978 | ||
| XM_006253070.2 | XP_006253132.1 | 7 | 1134 | |||
| XM_006253071.2 | XP_006253133.1 | 6 | 1082 | |||
| XM_006253073.2 | XP_006253135.1 | 6 | 4058 | |||
| XM_006253075.2 | XP_006253137.1 | 6 | 1082 | |||
| XM_006253076.2 | XP_006253138.1 | 6 | 1974 | |||
| XM_006253077.2 | XP_006253139.1 | 6 | 1080 | |||
| GenBank mRNA | AF142778.1 | - | 7 | 978 | ||
| D17521.1 | - | 6 | 1097 | |||

Clcn3
Genomic Map