siRNA Details
| Gene Symbol: Clcn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053363.2 | NP_445815.2 | 6 | 879 | ||
| XM_006253070.2 | XP_006253132.1 | 6 | 1035 | |||
| XM_006253071.2 | XP_006253133.1 | 5 | 983 | |||
| XM_006253073.2 | XP_006253135.1 | 5 | 3959 | |||
| XM_006253075.2 | XP_006253137.1 | 5 | 983 | |||
| XM_006253076.2 | XP_006253138.1 | 5 | 1875 | |||
| XM_006253077.2 | XP_006253139.1 | 5 | 981 | |||
| GenBank mRNA | AF142778.1 | - | 6 | 879 | ||
| D17521.1 | - | 5 | 998 | |||

Clcn3
Genomic Map