siRNA Details
| Gene Symbol: Prl7d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053364.1 | NP_445816.1 | 2 | 175 | ||
| GenBank mRNA | AF139809.1 | - | 2 | 175 | ||
| AF226609.1 | - | 2 | 147 | |||
| FQ219949.1 | - | 2 | 171 | |||
| FQ220120.1 | - | 2 | 122 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prl7d1| Gene Symbol: Prl7d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053364.1 | NP_445816.1 | 2 | 175 | ||
| GenBank mRNA | AF139809.1 | - | 2 | 175 | ||
| AF226609.1 | - | 2 | 147 | |||
| FQ219949.1 | - | 2 | 171 | |||
| FQ220120.1 | - | 2 | 122 | |||