siRNA Details
| Gene Symbol: Dhh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053367.1 | NP_445819.1 | 3 | 1444 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dhh| Gene Symbol: Dhh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053367.1 | NP_445819.1 | 3 | 1444 | ||