siRNA Details
| Gene Symbol: Tcf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053369.1 | NP_445821.1 | 12 | 1192 | ||
| GenBank mRNA | AF149284.1 | - | 12 | 1192 | ||
| FQ221525.1 | - | 943 | ||||
| FQ226283.1 | - | 3 | 331 | |||
| FQ233045.1 | - | 8, 9 | 878 | |||
| U09228.1 | - | 6 | 599 | |||

Tcf4
Genomic Map