siRNA Details
| Gene Symbol: Hcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053375.1 | NP_445827.1 | 8 | 2184 | ||
| GenBank mRNA | AF247450.1 | - | 8 | 2184 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hcn1| Gene Symbol: Hcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053375.1 | NP_445827.1 | 8 | 2184 | ||
| GenBank mRNA | AF247450.1 | - | 8 | 2184 | ||