siRNA Details
| Gene Symbol: Ihh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053384.1 | NP_445836.1 | 3 | 1065 | ||
| GenBank mRNA | AF175209.1 | - | 1 | 189 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ihh| Gene Symbol: Ihh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053384.1 | NP_445836.1 | 3 | 1065 | ||
| GenBank mRNA | AF175209.1 | - | 1 | 189 | ||