siRNA Details
| Gene Symbol: Ihh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053384.1 | NP_445836.1 | 3 | 952 | ||
| GenBank mRNA | AF162914.1 | - | 2 | 421 | ||
| AF175209.1 | - | 1 | 76 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ihh| Gene Symbol: Ihh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053384.1 | NP_445836.1 | 3 | 952 | ||
| GenBank mRNA | AF162914.1 | - | 2 | 421 | ||
| AF175209.1 | - | 1 | 76 | |||