siRNA Details
| Gene Symbol: Prelp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053385.1 | NP_445837.1 | 2 | 612 | ||
| XM_006249890.2 | XP_006249952.1 | 2 | 866 | |||
| XM_006249891.2 | XP_006249953.1 | 3 | 1210 | |||
| XM_008769571.1 | XP_008767793.1 | 2 | 880 | |||
| XM_008769572.1 | XP_008767794.1 | 3 | 1225 | |||
| XM_008769573.1 | XP_008767795.1 | 2 | 967 | |||
| GenBank mRNA | AF163569.1 | - | 2 | 612 | ||
| BC072487.1 | - | 2 | 630 | 7104853 | ||

Prelp
Genomic Map