siRNA Details
| Gene Symbol: Cdh11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053392.1 | NP_445844.1 | 8 | 1578 | ||
| XM_006255059.2 | XP_006255121.1 | 8 | 3658 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdh11| Gene Symbol: Cdh11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053392.1 | NP_445844.1 | 8 | 1578 | ||
| XM_006255059.2 | XP_006255121.1 | 8 | 3658 | |||